Assay Details
| Gene Symbol: Pbk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079937.1 | NP_001073406.1 | 2 - 3 | 308 | 89 | |
| XM_006252102.1 | XP_006252164.1 | 2 - 3 | 189 | 89 | ||
| XM_006252103.2 | XP_006252165.1 | 2 - 3 | 167 | 89 | ||
| XM_006252104.2 | XP_006252166.1 | 2 - 3 | 178 | 89 | ||
| XM_006252105.2 | XP_006252167.1 | 2 - 3 | 447 | 89 | ||
| XM_006252106.2 | XP_006252168.1 | 2 - 3 | 387 | 89 | ||
| GenBank mRNA | BC129099.1 | - | 2 - 3 | 308 | 8375047 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Pbk
Genomic Map