Assay Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271488.1 | NP_001258417.1 | 4 - 5 | 654 | 74 | |
| XM_003751499.3 | XP_003751547.1 | 4 - 5 | 2296 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ints6
Genomic Map