Assay Details
| Gene Symbol: Sgcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006993.1 | NP_001006994.1 | 7 - 8 | 799 | 85 | |
| GenBank mRNA | BC083748.1 | - | 7 - 8 | 799 | 7192349 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104085 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14597 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14597 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14597 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14597 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31059 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64678 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63424 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104108 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784348 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784353 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784351 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784352 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784347 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784350 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784354 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784349 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784346 | - | - |
| Rat Genome 230 2.0 Array | 1374683_at | - | - |
| Rat Genome 230 2.0 Array | 1374683_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374683_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374683_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500012284.rn.1 | TC1500001832.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374683_PM_at | - | - |
| Rat Expression Set 230 | 1374683_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784346 | - | - |
| Rat Genome U34 Set | rc_AA850867_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369105 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68729 | - | - |

Sgcg
Genomic Map