Assay Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729770.4 | XP_002729816.1 | 1 - 2 | 2148 | 113 | |
| XM_008765188.1 | XP_008763410.1 | 2 - 3 | 290 | 113 | ||
| GenBank mRNA | FQ222140.1 | - | 1 - 2 | 120 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002729770.4, XM_008765188.1 transcripts.
Set Membership: |
|

Zfp952
Genomic Map