Assay Details
| Gene Symbol: Stk11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108069.1 | NP_001101539.1 | 7 - 8 | 1743 | 59 | |
| XM_006240910.1 | XP_006240972.1 | 7 - 8 | 1738 | 59 | ||
| XM_008765096.1 | XP_008763318.1 | 7 - 8 | 1738 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
tissue homeostasis
vasculature development
protein phosphorylation
autophagy
cellular response to DNA damage stimulus
cell cycle arrest
spermatid development
axonogenesis
negative regulation of cell proliferation
response to ionizing radiation
establishment of cell polarity
regulation of Wnt signaling pathway
negative regulation of cell growth
positive regulation of transforming growth factor beta receptor signaling pathway
activation of protein kinase activity
response to glucagon
response to lipid
TCR signalosome assembly
glucose homeostasis
anoikis
response to peptide hormone
positive thymic T cell selection
positive regulation of gluconeogenesis
positive regulation of protein kinase activity
protein autophosphorylation
regulation of dendrite morphogenesis
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of axonogenesis
T cell receptor signaling pathway
protein heterooligomerization
Golgi localization
regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of epithelial cell proliferation involved in prostate gland development
cellular response to UV-B
intrinsic apoptotic signaling pathway by p53 class mediator
dendrite extension
positive regulation of protein localization to nucleus
magnesium ion binding
p53 binding
protein serine/threonine kinase activity
ATP binding
LRR domain binding
protein kinase activator activity
protein complex binding
regulation of cell growth
tissue homeostasis
vasculature development
protein phosphorylation
autophagy
cellular response to DNA damage stimulus
cell cycle arrest
spermatid development
axonogenesis
negative regulation of cell proliferation
response to ionizing radiation
establishment of cell polarity
regulation of Wnt signaling pathway
negative regulation of cell growth
positive regulation of transforming growth factor beta receptor signaling pathway
activation of protein kinase activity
response to glucagon
response to lipid
TCR signalosome assembly
glucose homeostasis
anoikis
response to peptide hormone
positive thymic T cell selection
positive regulation of gluconeogenesis
positive regulation of protein kinase activity
protein autophosphorylation
regulation of dendrite morphogenesis
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of axonogenesis
T cell receptor signaling pathway
protein heterooligomerization
Golgi localization
regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of epithelial cell proliferation involved in prostate gland development
cellular response to UV-B
intrinsic apoptotic signaling pathway by p53 class mediator
dendrite extension
positive regulation of protein localization to nucleus
magnesium ion binding
p53 binding
protein serine/threonine kinase activity
ATP binding
LRR domain binding
protein kinase activator activity
protein complex binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P410670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 829 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P410670 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P410670 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P410670 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P410670 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P410676 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31672 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10900615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900611 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900614 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900616 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10900606 | - | - |
| Rat Genome 230 2.0 Array | 1381830_PM_x_at | - | - |
| Rat Genome 230 2.0 Array | 1375364_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375364_at | - | - |
| Rat Genome 230 2.0 Array | 1375238_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375238_at | - | - |
| Rat Genome 230 2.0 Array | 1381830_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1375364_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381830_PM_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1375238_at | - | - |
| Affymetrix HT Rat Focus Array | 1375364_at | - | - |
| Affymetrix HT Rat Focus Array | 1375238_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381830_x_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700017310.rn.1 | TC0700002177.rn.1 | RTA TC |
| Rat Expression Set 230 | 1375364_PM_at | - | - |
| Rat Expression Set 230 | 1375238_PM_at | - | - |
| Rat Expression Set 230 | 1375364_at | - | - |
| Rat Expression Set 230 | 1375238_at | - | - |
| Rat Expression Set 230 | 1381830_x_at | - | - |
| Rat Expression Set 230 | 1381830_PM_x_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10900606 | - | - |
| Rat Genome U34 Set | rc_AI045508_at | - | - |
| Rat Genome U34 Set | rc_AI113337_at | - | - |
| Rat Genome U34 Set | rc_AI136725_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21645566 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366315 | - | - |

Stk11
Genomic Map