Assay Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014762.1 | NP_001014762.1 | 2 - 3 | 192 | 67 | |
| GenBank mRNA | BC082021.1 | - | 2 - 3 | 192 | 7129610 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Pdrg1
Genomic Map