Assay Details
| Gene Symbol: LOC100911769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762428.1 | XP_008760650.1 | 4 - 5 | 578 | 108 | |
| XM_008762429.1 | XP_008760651.1 | 4 - 5 | 575 | 108 | ||
| XM_008762430.1 | XP_008760652.1 | 4 - 5 | 4020 | 108 | ||
| XM_008762431.1 | XP_008760653.1 | 4 - 5 | 578 | 108 | ||
| XM_008762432.1 | XP_008760654.1 | 4 - 5 | 578 | 108 | ||
| XM_008762433.1 | XP_008760655.1 | 4 - 5 | 578 | 108 | ||
| XM_008762434.1 | XP_008760656.1 | 4 - 5 | 578 | 108 | ||
| XM_008762435.1 | XP_008760657.1 | 4 - 5 | 578 | 108 | ||
| Gene Symbol: Epb41l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021681.2 | NP_067713.2 | 4 - 5 | 608 | 108 | |
| NM_172090.3 | NP_742087.1 | 4 - 5 | 591 | 108 | ||
| XM_008762381.1 | XP_008760603.1 | 4 - 5 | 608 | 108 | ||
| GenBank mRNA | AB019256.1 | - | 4 - 5 | 589 | 108 | |
| AB019257.1 | - | 4 - 5 | 608 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911769
Genomic Map