Assay Details
| Gene Symbol: Tpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107790.1 | NP_001101260.1 | 9 - 10 | 1443 | 72 | |
| XM_006235270.2 | XP_006235332.1 | 10 - 11 | 1346 | 72 | ||
| XM_006235272.2 | XP_006235334.1 | 10 - 11 | 1298 | 72 | ||
| XM_006235273.2 | XP_006235335.1 | 9 - 10 | 1228 | 72 | ||
| XM_006235274.2 | XP_006235336.1 | 10 - 11 | 1346 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpx2
Genomic Map