Assay Details
| Gene Symbol: Efcab8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762400.1 | XP_008760622.1 | 2 - 3 | 883 | 82 | |
| XM_008762402.1 | XP_008760624.1 | 2 - 3 | 884 | 82 | ||
More Information
Set Membership: |
|

Efcab8
Genomic Map