Assay Details
| Gene Symbol: Tbc1d20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004281.1 | NP_001004281.1 | 2 - 3 | 314 | 73 | |
| XM_008762348.1 | XP_008760570.1 | 2 - 3 | 463 | 73 | ||
| GenBank mRNA | BC079383.1 | - | 2 - 3 | 314 | 7111239 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d20
Genomic Map