Assay Details
| Gene Symbol: LOC100910990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749608.3 | XP_003749656.1 | 10 - 11 | 1022 | 58 | |
| XM_006235456.2 | XP_006235518.1 | 11 - 12 | 1104 | 58 | ||
| XM_006235457.2 | XP_006235519.1 | 11 - 12 | 1072 | 58 | ||
| XM_008762413.1 | XP_008760635.1 | 10 - 11 | 966 | 58 | ||
| XM_008762414.1 | XP_008760636.1 | 11 - 12 | 6180 | 58 | ||
| Gene Symbol: Cpne1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256465.1 | NP_001243394.1 | 856 | 58 | ||
| GenBank mRNA | AM747279.1 | - | 856 | 58 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910990
Genomic Map