Assay Details
| Gene Symbol: Procr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025733.2 | NP_001020904.1 | 1 - 2 | 197 | 53 | |
| GenBank mRNA | BC097380.1 | - | 1 - 2 | 168 | 7445753 | 53 |
| BC099115.1 | - | 1 - 2 | 197 | 7462042 | 53 | |
| FQ220150.1 | - | 1 - 2 | 236 | 53 | ||
| FQ221148.1 | - | 1 - 2 | 204 | 53 | ||
| FQ228484.1 | - | 1 - 2 | 202 | 53 | ||
| FQ229445.1 | - | 1 - 2 | 234 | 53 | ||
| FQ229483.1 | - | 1 - 2 | 204 | 53 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Procr
Genomic Map