Assay Details
| Gene Symbol: Dync1li2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031026.1 | NP_112288.1 | 2 - 3 | 189 | 72 | |
| XM_006255053.2 | XP_006255115.1 | 2 - 3 | 403 | 72 | ||
| XM_006255054.2 | XP_006255116.1 | 2 - 3 | 403 | 72 | ||
| GenBank mRNA | BC089794.1 | - | 2 - 3 | 210 | 7115029 | 72 |
| U15138.1 | - | 2 - 3 | 189 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dync1li2
Genomic Map