Assay Details
| Gene Symbol: Gpr97 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255126.2 | XP_006255188.1 | 8 - 9 | 2896 | 91 | |
| XM_006255127.2 | XP_006255189.1 | 7 - 8 | 2749 | 91 | ||
More Information
Set Membership: |
|

Gpr97
Genomic Map