Assay Details
| Gene Symbol: Mt2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137564.1 | NP_001131036.1 | 1 - 2 | 92 | 87 | |
| GenBank mRNA | BC168768.1 | - | 1 - 2 | 92 | 6891160 | 87 |
| FQ210821.1 | - | 1 - 2 | 95 | 87 | ||
| FQ211084.1 | - | 1 - 2 | 92 | 87 | ||
| FQ218212.1 | - | 1 - 2 | 92 | 87 | ||
| FQ218903.1 | - | 1 - 2 | 92 | 87 | ||
| FQ219667.1 | - | 1 - 2 | 92 | 87 | ||
| FQ221583.1 | - | 1 - 2 | 89 | 87 | ||
| FQ222390.1 | - | 1 - 2 | 92 | 87 | ||
| FQ222722.1 | - | 1 - 2 | 92 | 87 | ||
| FQ222943.1 | - | 1 - 2 | 94 | 87 | ||
| FQ228591.1 | - | 1 - 2 | 92 | 87 | ||
| FQ228662.1 | - | 1 - 2 | 92 | 87 | ||
| FQ229013.1 | - | 1 - 2 | 92 | 87 | ||
| Gene Symbol: Mt1m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728917.3 | XP_002728963.1 | 1 | 98 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC168768.1, NM_001137564.1, XM_002728917.3 transcripts.
Set Membership: |
|

Mt2A
Genomic Map