Assay Details
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106165.1 | NP_001099635.1 | 2 - 3 | 335 | 69 | |
| XM_006255051.2 | XP_006255113.1 | 2 - 3 | 463 | 69 | ||
| XM_008772243.1 | XP_008770465.1 | 2 - 3 | 297 | 69 | ||
| GenBank mRNA | BC167038.1 | - | 2 - 3 | 304 | 9090629 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Car7
Genomic Map