Assay Details
| Gene Symbol: Car7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106165.1 | NP_001099635.1 | 3 - 4 | 455 | 65 | |
| XM_006255051.2 | XP_006255113.1 | 3 - 4 | 583 | 65 | ||
| GenBank mRNA | BC167038.1 | - | 3 - 4 | 424 | 9090629 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Car7
Genomic Map