Assay Details
| Gene Symbol: Cklf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139111.1 | NP_620811.1 | 2 - 3 | 401 | 182 | |
| GenBank mRNA | AF253065.2 | - | 2 - 3 | 401 | 182 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cklf
Genomic Map