Assay Details
| Gene Symbol: Polr2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012473.1 | NP_001012491.1 | 4 - 5 | 281 | 68 | |
| GenBank mRNA | BC089905.1 | - | 4 - 5 | 281 | 7322752 | 68 |
| FQ218345.1 | - | 4 - 5 | 198 | 68 | ||
| FQ225242.1 | - | 4 - 5 | 301 | 68 | ||
| FQ226282.1 | - | 4 - 5 | 320 | 68 | ||
| FQ227427.1 | - | 4 - 5 | 301 | 68 | ||
| FQ235323.1 | - | 4 - 5 | 320 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Polr2c
Genomic Map