Assay Details
| Gene Symbol: LOC103689945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770640.1 | XP_008768862.1 | 11 - 12 | 2165 | 93 | |
| Gene Symbol: Zswim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271275.1 | NP_001258204.1 | 12 - 13 | 2630 | 93 | |
| XM_008770546.1 | XP_008768768.1 | 11 - 12 | 2744 | 93 | ||
| XM_008770548.1 | XP_008768770.1 | 11 - 12 | 2743 | 93 | ||
| XM_008770550.1 | XP_008768772.1 | 12 - 13 | 2642 | 93 | ||
| XM_008770551.1 | XP_008768773.1 | 11 - 12 | 2744 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103689945
Genomic Map