Assay Details
| Gene Symbol: LOC100909469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751425.3 | XP_003751473.2 | 3 - 4 | 466 | 77 | |
| XM_008770474.1 | XP_008768696.1 | 3 - 4 | 1060 | 77 | ||
| Gene Symbol: Dusp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162408.1 | NP_001155880.1 | 899 | 77 | ||
| XM_008770542.1 | XP_008768764.1 | 3 - 4 | 1060 | 77 | ||
| GenBank mRNA | BC166532.1 | - | 899 | 7190084 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909469
Genomic Map