Assay Details
| Gene Symbol: Tppp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227793.2 | XP_006227855.1 | 4 | 5337 | 83 | |
| XM_006227794.2 | XP_006227856.1 | 4 | 5268 | 83 | ||
| GenBank mRNA | FQ212396.1 | - | 1 | 1062 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tppp
Genomic Map