Assay Details
| Gene Symbol: Papd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107333.1 | NP_001100803.1 | 2 - 3 | 303 | 78 | |
| XM_006227781.2 | XP_006227843.1 | 2 - 3 | 4053 | 78 | ||
| XM_006227782.2 | XP_006227844.1 | 2 - 3 | 690 | 78 | ||
| XM_008758690.1 | XP_008756912.1 | 2 - 3 | 420 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papd7
Genomic Map