Assay Details
| Gene Symbol: Tppp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108461.1 | NP_001101931.1 | 2 - 3 | 389 | 71 | |
| XM_006227793.2 | XP_006227855.1 | 2 - 3 | 630 | 71 | ||
| XM_006227794.2 | XP_006227856.1 | 2 - 3 | 561 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tppp
Genomic Map