Assay Details
| Gene Symbol: Exoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024964.1 | NP_001020135.1 | 12 - 13 | 2141 | 63 | |
| XM_006227767.2 | XP_006227829.1 | 12 - 13 | 2259 | 63 | ||
| XM_006227768.2 | XP_006227830.1 | 12 - 13 | 2555 | 63 | ||
| GenBank mRNA | BC097993.1 | - | 12 - 13 | 2141 | 7446619 | 63 |
| U32575.1 | - | 11 - 12 | 2096 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P120971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51193 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125531 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P120971 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P120971 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P120971 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P381837 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700912362 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91517 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74972 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700912362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10702457 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702458 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702460 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702464 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702465 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702455 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702466 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702462 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702453 | - | - |
| Rat Genome 230 2.0 Array | 1374656_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380746_at | - | - |
| Rat Genome 230 2.0 Array | 1374656_at | - | - |
| Rat Genome 230 2.0 Array | 1370686_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370686_at | - | - |
| Rat Genome 230 2.0 Array | 1389624_s_at | - | - |
| Rat Genome 230 2.0 Array | 1380746_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389624_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1374656_at | - | - |
| Affymetrix HT Rat Focus Array | 1389624_PM_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1370686_at | - | - |
| Affymetrix HT Rat Focus Array | 1374656_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370686_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389624_s_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100001913.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001910.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001908.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001909.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001921.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001920.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001905.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001911.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001914.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001916.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001918.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001912.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001915.rn.1 | TC0100000331.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001917.rn.1 | TC0100000331.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374656_PM_at | - | - |
| Rat Expression Set 230 | 1389624_PM_s_at | - | - |
| Rat Expression Set 230 | 1389624_s_at | - | - |
| Rat Expression Set 230 | 1374656_at | - | - |
| Rat Expression Set 230 | 1370686_at | - | - |
| Rat Expression Set 230 | 1370686_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702453 | - | - |
| Rat Genome U34 Set | U32575_at | - | - |
| Rat Genome U34 Set | U32575_g_at | - | - |
| Rat Genome U34 Set | rc_AI045764_at | - | - |
| Rat Genome U34 Set | rc_AA851443_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373269 | - | - |

Exoc3
Genomic Map