Assay Details
| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100990.1 | NP_001094460.1 | 2 - 3 | 391 | 152 | |
| GenBank mRNA | EU030376.1 | - | 2 - 3 | 391 | 152 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rspo3
Genomic Map