Assay Details
| Gene Symbol: Rspo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100990.1 | NP_001094460.1 | 3 - 4 | 533 | 76 | |
| GenBank mRNA | EU030376.1 | - | 3 - 4 | 533 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
biological_process
canonical Wnt signaling pathway
branching involved in labyrinthine layer morphogenesis
positive regulation of canonical Wnt signaling pathway
G-protein coupled receptor binding
molecular_function
receptor binding
frizzled binding
heparin binding
angiogenesis
biological_process
canonical Wnt signaling pathway
branching involved in labyrinthine layer morphogenesis
positive regulation of canonical Wnt signaling pathway
G-protein coupled receptor binding
molecular_function
receptor binding
frizzled binding
heparin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P22653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20664 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P22653 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P22653 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P22653 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22653 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17026 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22352 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10702417 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702414 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702416 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702415 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702412 | - | - |
| Rat Genome 230 2.0 Array | 1396544_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1396544_at | - | - |
| Rat Genome 230 2.0 Array | 1383873_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383873_at | - | - |
| Rat Genome 230 2.0 Array | 1390632_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390632_at | - | - |
| Affymetrix HT Rat Focus Array | 1396544_at | - | - |
| Affymetrix HT Rat Focus Array | 1383873_at | - | - |
| Affymetrix HT Rat Focus Array | 1383873_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1396544_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100001759.rn.1 | TC0100000311.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001761.rn.1 | TC0100000311.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001762.rn.1 | TC0100000311.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100001760.rn.1 | TC0100000311.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390632_PM_at | - | - |
| Rat Expression Set 230 | 1390632_at | - | - |
| Rat Expression Set 230 | 1396544_PM_at | - | - |
| Rat Expression Set 230 | 1396544_at | - | - |
| Rat Expression Set 230 | 1383873_PM_at | - | - |
| Rat Expression Set 230 | 1383873_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702412 | - | - |
| Rat Genome U34 Set | rc_AI045979_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20949478 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363395 | - | - |

Rspo3
Genomic Map