Assay Details
| Gene Symbol: LOC100910446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758670.1 | XP_008756892.1 | 1 - 2 | 135 | 112 | |
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021869.2 | NP_068641.2 | 3 - 4 | 249 | 112 | |
| XM_006227691.2 | XP_006227753.1 | 4 - 5 | 1571 | 112 | ||
| XM_008758626.1 | XP_008756848.1 | 4 - 5 | 1565 | 112 | ||
| GenBank mRNA | AF031430.1 | - | 2 - 3 | 153 | 112 | |
| BC085737.1 | - | 3 - 4 | 249 | 7193349 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100910446
Genomic Map