Assay Details
| Gene Symbol: Slc12a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013144.2 | NP_001013162.2 | 4 - 5 | 536 | 72 | |
| XM_006227789.2 | XP_006227851.1 | 4 - 5 | 661 | 72 | ||
| GenBank mRNA | BC086339.1 | - | 4 - 5 | 535 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a7
Genomic Map