Assay Details
| Gene Symbol: Eya4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271317.1 | NP_001258246.1 | 11 - 12 | 1220 | 62 | |
| XM_008758600.1 | XP_008756822.1 | 11 - 12 | 1082 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eya4
Genomic Map