Assay Details
| Gene Symbol: Rpp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108372.2 | NP_001101842.2 | 3 - 4 | 432 | 101 | |
| NM_001141929.1 | NP_001135401.1 | 3 - 4 | 387 | 101 | ||
| XM_008770552.1 | XP_008768774.1 | 3 - 4 | 656 | 101 | ||
| GenBank mRNA | BU670982.1 | - | 3 - 4 | 432 | 101 | |
| CK356697.1 | - | 3 - 4 | 387 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpp14
Genomic Map