Assay Details
| Gene Symbol: Nr1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147210.2 | NP_671743.2 | 7 - 8 | 1742 | 86 | |
| GenBank mRNA | U15660.1 | - | 6 - 7 | 1459 | 86 | |
| U20796.1 | - | 6 - 7 | 1399 | 86 | ||
| X82777.1 | - | 7 - 8 | 1677 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr1d2
Genomic Map