Assay Details
| Gene Symbol: Cyld-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008763560.1 | XP_008761782.1 | 1 | 2143 | 114 | |
| Gene Symbol: Cyld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017380.1 | NP_001017380.1 | 10 - 11 | 2130 | 114 | |
| XM_008772408.1 | XP_008770630.1 | 10 - 11 | 3379 | 114 | ||
| GenBank mRNA | BC082001.1 | - | 10 - 11 | 2130 | 114 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cyld-ps1
Genomic Map