Assay Details
| Gene Symbol: Papd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107416.1 | NP_001100886.1 | 3 - 4 | 425 | 118 | |
| XM_006255261.1 | XP_006255323.1 | 4 - 5 | 906 | 118 | ||
| XM_006255263.2 | XP_006255325.1 | 4 - 5 | 906 | 118 | ||
| XM_008772403.1 | XP_008770625.1 | 5 - 6 | 917 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Papd5
Genomic Map