Assay Details
| Gene Symbol: Papd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107416.1 | NP_001100886.1 | 9 - 10 | 1335 | 59 | |
| XM_006255261.1 | XP_006255323.1 | 11 - 12 | 1945 | 59 | ||
| XM_006255263.2 | XP_006255325.1 | 11 - 12 | 1945 | 59 | ||
| XM_008772403.1 | XP_008770625.1 | 12 - 13 | 1956 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P051297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143215 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051297 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P051297 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P051297 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P535555 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P142606 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83457 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63183 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10809718 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809717 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809722 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809724 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10809716 | - | - |
| Rat Genome 230 2.0 Array | 1374612_at | - | - |
| Rat Genome 230 2.0 Array | 1374612_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374612_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374612_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900008857.rn.1 | TC1900001100.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008867.rn.1 | TC1900001100.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008866.rn.1 | TC1900001100.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008858.rn.1 | TC1900001100.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008860.rn.1 | TC1900001100.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008865.rn.1 | TC1900001100.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374612_PM_at | - | - |
| Rat Expression Set 230 | 1374612_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809716 | - | - |
| Rat Genome U34 Set | rc_AI170664_at | - | - |
| Rat Genome U34 Set | rc_AI072670_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21906904 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1651034 | - | - |

Papd5
Genomic Map