Assay Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253993.2 | XP_006254055.1 | 1 - 2 | 202 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006253993.2 transcripts.
Set Membership: |
|

Rala
Genomic Map