Assay Details
| Gene Symbol: LOC100912163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771836.1 | XP_008770058.1 | 3 - 4 | 303 | 85 | |
| Gene Symbol: Arid4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053421.1 | NP_445873.1 | 3 - 4 | 218 | 85 | |
| XM_006254016.2 | XP_006254078.1 | 4 - 5 | 543 | 85 | ||
| XM_006254019.2 | XP_006254081.1 | 4 - 5 | 543 | 85 | ||
| XM_006254020.2 | XP_006254082.1 | 4 - 5 | 543 | 85 | ||
| XM_008771764.1 | XP_008769986.1 | 4 - 5 | 543 | 85 | ||
| XM_008771765.1 | XP_008769987.1 | 4 - 5 | 543 | 85 | ||
| XM_008771766.1 | XP_008769988.1 | 4 - 5 | 543 | 85 | ||
| GenBank mRNA | AF245512.1 | - | 3 - 4 | 218 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100912163
Genomic Map