Assay Details
| Gene Symbol: Blnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025767.1 | NP_001020938.1 | 16 - 17 | 1363 | 96 | |
| GenBank mRNA | BC098790.1 | - | 16 - 17 | 1363 | 7375523 | 96 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P144741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160714 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P144741 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P144741 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P144741 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P144741 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67748 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101922 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1352 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57200 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730142 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730147 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730146 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730144 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730141 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730145 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730143 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730140 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730131 | - | - |
| Rat Genome 230 2.0 Array | 1393641_at | - | - |
| Rat Genome 230 2.0 Array | 1393641_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393641_at | - | - |
| Affymetrix HT Rat Focus Array | 1393641_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100061762.rn.1 | TC0100007686.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393641_PM_at | - | - |
| Rat Expression Set 230 | 1393641_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730131 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21591819 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1354453 | - | - |

Blnk
Genomic Map