Assay Details
| Gene Symbol: Igfbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012588.2 | NP_036720.2 | 1 - 2 | 506 | 103 | |
| GenBank mRNA | M31837.1 | - | 1 - 2 | 459 | 103 | |
| M33300.1 | - | 1 - 2 | 436 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Igfbp3
Genomic Map