Assay Details
| Gene Symbol: Ctsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013156.2 | NP_037288.1 | 5 - 6 | 692 | 95 | |
| GenBank mRNA | AB630323.1 | - | 4 - 5 | 633 | 95 | |
| BC063175.1 | - | 5 - 6 | 707 | 6918389 | 95 | |
| FQ209613.1 | - | 5 - 6 | 703 | 95 | ||
| FQ218649.1 | - | 5 - 6 | 701 | 95 | ||
| FQ228725.1 | - | 5 - 6 | 703 | 95 | ||
| FQ228844.1 | - | 5 - 6 | 703 | 95 | ||
| Y00697.1 | - | 5 - 6 | 696 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsl
Genomic Map