Assay Details
| Gene Symbol: Syt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031666.1 | NP_113854.1 | 6 - 7 | 1494 | 98 | |
| GenBank mRNA | AF375463.1 | - | 6 - 7 | 1494 | 98 | |
| DQ437524.1 | - | 6 - 7 | 1494 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P379342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121696 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163062 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P379342 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P379342 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P379342 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P379342 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60474 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99334 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10906366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906369 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906365 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906367 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906370 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906368 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906371 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10906364 | - | - |
| Rat Genome 230 2.0 Array | 1370718_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370718_at | - | - |
| Affymetrix HT Rat Focus Array | 1370718_at | - | - |
| Affymetrix HT Rat Focus Array | 1370718_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700029656.rn.1 | TC0700003711.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370718_PM_at | - | - |
| Rat Expression Set 230 | 1370718_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10906364 | - | - |
| Rat Genome U34 Set | U85513_at | - | - |
| Rat Genome U34 Set | U85513_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21846461 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360193 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60668 | - | - |

Syt10
Genomic Map