Assay Details
| Gene Symbol: Cd40lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053353.1 | NP_445805.1 | 1 - 2 | 159 | 150 | |
| GenBank mRNA | AF013985.1 | - | 1 - 2 | 159 | 150 | |
| AF116582.1 | - | 1 - 2 | 159 | 150 | ||
| EF066490.1 | - | 1 - 2 | 159 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd40lg
Genomic Map