Assay Details
| Gene Symbol: Slc35a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147140.2 | NP_671481.2 | 2 | 1768 | 75 | |
| XM_006254528.1 | XP_006254590.1 | 2 | 1802 | 75 | ||
| XM_006254529.2 | XP_006254591.1 | 4 | 1634 | 75 | ||
| XM_008772019.1 | XP_008770241.1 | 2 | 1721 | 75 | ||
| GenBank mRNA | AF406814.1 | - | 1 | 1539 | 75 | |
| BC085695.1 | - | 2 | 1768 | 7191954 | 75 | |
| BC086986.1 | - | 2 | 1721 | 7192444 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc35a4
Genomic Map