Assay Details
| Gene Symbol: Yrdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175604.2 | NP_783194.2 | 2 - 3 | 524 | 71 | |
| GenBank mRNA | AY172974.1 | - | 250 | 71 | ||
| BC088428.1 | - | 2 - 3 | 489 | 71 | ||
| BC099797.1 | - | 2 - 3 | 524 | 7936110 | 71 | |
More Information
Set Membership: |
|

Yrdc
Genomic Map