Assay Details
| Gene Symbol: Impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199099.2 | NP_954530.1 | 6 - 7 | 670 | 84 | |
| XM_006243751.2 | XP_006243813.1 | 6 - 7 | 711 | 84 | ||
| GenBank mRNA | BC060585.1 | - | 6 - 7 | 671 | 6891123 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Impdh2
Genomic Map