Assay Details
| Gene Symbol: Xirp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201989.1 | NP_973718.1 | 8 - 9 | 10183 | 69 | |
| GenBank mRNA | AF466694.1 | - | 8 - 9 | 10183 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P231223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126171 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P231223 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P231223 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P231223 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P231223 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10264 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71383 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66591 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10836506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10836505 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10836504 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300005388.rn.1 | TC0300000721.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300005390.rn.1 | TC0300000721.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10836504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20927247 | - | - |

Xirp2
Genomic Map