Assay Details
| Gene Symbol: Atf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012912.2 | NP_037044.1 | 3 - 4 | 505 | 88 | |
| GenBank mRNA | BC078903.1 | - | 3 - 4 | 505 | 7100767 | 88 |
| M63282.1 | - | 3 - 4 | 510 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atf3
Genomic Map