Assay Details
| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013053.1 | NP_037185.1 | 1 - 2 | 47 | 94 | |
| GenBank mRNA | D17614.1 | - | 1 - 2 | 47 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the D17614.1, NM_013053.1 transcripts.
Set Membership: |
|

Ywhaq
Genomic Map