Assay Details
| Gene Symbol: LOC100909595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749833.3 | XP_003749881.1 | 1 - 2 | 350 | 121 | |
| Gene Symbol: Slc2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC063168.1 | - | 1 - 2 | 143 | 6887820 | 121 |
| D13962.1 | - | 100 | 121 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC063168.1, D13962.1, XM_003749833.3 transcripts.
Set Membership: |
|

LOC100909595
Genomic Map