Assay Details
| Gene Symbol: Alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017260.2 | NP_058956.1 | 4 - 5 | 375 | 62 | |
| GenBank mRNA | BC086593.1 | - | 4 - 5 | 375 | 7310271 | 62 |
| X52196.1 | - | 4 - 5 | 357 | 62 | ||
More Information
Set Membership: |
|

Alox5ap
Genomic Map